missing translation for 'onlineSavingsMsg'
Learn More

Econazole nitrate

Product Code. 15485439
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485439 25 g 25g
15475439 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15485439 Supplier Thermo Scientific Alfa Aesar Supplier No. J63173.14
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 24169-02-6 | C18H16Cl3N3O4 | 444.693 g/mol

Econazole nitrate is an antifungal medication often used as a cream to treat skin infections like tinea, ringworm and athlete's foot, personal care, cosmetic and pharmaceutical applications. It is used for studies on processes such as platelet serotonin uptake, prostanoid biosynthesis, EDHF-mediated relaxation, and Ca2+ transport pathways in thymic lymphocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24169-02-6
Molecular Formula C18H16Cl3N3O4
Molecular Weight (g/mol) 444.693
MDL Number MFCD00058160
InChI Key DDXORDQKGIZAME-UHFFFAOYSA-N
Synonym econazole nitrate, ifenec, epi-pevaryl, gyno-pevaryl, econazole nitrate salt, pevaryl, +--econazole nitrate, spectazole, econazolum nitrate, gyno-pevaryl 150
PubChem CID 68589
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
SMILES C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]

Specifications

Melting Point 158°C to 163°C
Packaging Plastic bottle
Quantity 25 g
Solubility Information Soluble at 0.4mg/ml in DMSO. Sparingly soluble in water.
Formula Weight 444.7
Physical Form Powder
Chemical Name or Material Econazole nitrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.