missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Econazole, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Econazole |
| CAS | 27220-47-9 |
| Analyte or Component Names | Econazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H15 Cl3 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Econazole, 1-[(2RS)-2-[(4-Chlorobenzyl)oxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, 1H-Imidazole, 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-, NSC 187789, Imidazole, 1-[2,4-dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]- (8CI), (+/-)-Econazole, 1-[2,4-Dichloro-beta-[(p-chlorobenzyl)oxy]phenethyl]imidazole |
| InChI Formula | InChI=1S/C18H15Cl3N2O/c19-14-3-1-13(2-4-14)11-24-18(10-23-8-7-22-12-23)16-6-5-15(20)9-17(16)21/h1-9,12,18H,10-11H2 |
| Solution Type | Neat |
| SMILES | Clc1ccc(COC(Cn2ccnc2)c3ccc(Cl)cc3Cl)cc1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction