missing translation for 'onlineSavingsMsg'
Learn More

Econazole, 99%, Thermo Scientific Chemicals

Product Code. p-7077745 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15540584

Brand: Thermo Scientific Acros 458630050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27220-47-9
Molecular Formula C18H15Cl3N2O
Molecular Weight (g/mol) 381.68
MDL Number MFCD00800993,MFCD00058160 (.HNO3)
InChI Key LEZWWPYKPKIXLL-UHFFFAOYNA-N
Synonym econazole, ecostatin, econazolum, pevaryl, gyno-pevaryl, econazol, palavale, spectazole, +--econazole, ecostatin cream
PubChem CID 3198
ChEBI CHEBI:82873
IUPAC Name 1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole
SMILES ClC1=CC=C(COC(CN2C=CN=C2)C2=C(Cl)C=C(Cl)C=C2)C=C1

Tekniske data

Melting Point 91.0°C to 94.0°C
Loss on Drying 0.5% max. (1 g, 60°C, 4 h)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 5 g
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in hot methanol, slightly soluble in ethanol, insoluble in ether
Formula Weight 381.68
Percent Purity 99%
Chemical Name or Material Econazole
compliance-icons
Produktidentifikator
  • Econazole
Signalord
  • Warning
Farekategori
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.