missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eco1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-84836-0.1ML
This item is not returnable.
View return policy
Description
Eco1 Polyclonal specifically detects Eco1 in Human samples. It is validated for Western Blot.
Specifications
| Eco1 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| CTF7 homolog 1, EC 2.3.1.-, ECO1 homolog 1, EFO1CTF, EFO1p, ESO1 homolog 1, ESO1A930014I12Rik, Establishment factor-like protein 1, Establishment of cohesion 1 homolog 1, establishment of cohesion 1 homolog 1 (S. cerevisiae), hEFO1, KIAA1911ECO1, MGC105022, N-acetyltransferase ESCO1 | |
| The immunogen is a synthetic peptide directed towards the N-terminal region of Eco1 | |
| 0.1 mL | |
| DNA Repair, Lipid and Metabolism | |
| 114799 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction