missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eco1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-87957
This item is not returnable.
View return policy
Description
Eco1 Polyclonal specifically detects Eco1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Eco1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| CTF7 homolog 1, EC 2.3.1.-, ECO1 homolog 1, EFO1CTF, EFO1p, ESO1 homolog 1, ESO1A930014I12Rik, Establishment factor-like protein 1, Establishment of cohesion 1 homolog 1, establishment of cohesion 1 homolog 1 (S. cerevisiae), hEFO1, KIAA1911ECO1, MGC105022, N-acetyltransferase ESCO1 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ESCO1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ERAPENCHLANEIKPSDPPLDNQMKHSFDSASNKNFSQCLESKLENSPVENVTAASTLLSQAKIDTGENKFPGSAPQQHSILSNQTSKSSDNRETPRNHSLPKCNSHLEI | |
| 0.1 mL | |
| DNA Repair, Lipid and Metabolism | |
| 114799 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction