missing translation for 'onlineSavingsMsg'
Learn More

SHOWA™ Eco Best Technology™ Disposable Nitrile Gloves

Product Code. 17094367
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17094367 Small Pack of 100
17857311 X-Small Pack of 100
17851181 Medium Pack of 100
17861181 Large Pack of 100
17871181 X-Large Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 17094367 Supplier SHOWA™ Supplier No. 6112PF07S
Only null left
Add to basket
Add to basket

Eco Best Technology™ (EBT) biodegradable nitrile disposable gloves provide comfort, dexterity and performance across a wide range of industrial applications.

  • Same performance as non-EBT nitrile disposable gloves
  • Eco Best Technology™ (EBT) achieved 82% biodegradation in 386 days
  • Decreased risk of allergies
  • Second-skin fit and feel
  • Recommended for industrial applications including food, pharmaceutical, utilities, construction, agricultural and automotive

Compliance

Data Testing is available upon request.

Specifications

Certifications/Compliance ASTM D5511-18, ASTM D5526
Certifications/Compliance (Europe) EN 374-2:2003, EN 374-1:2016/Type B, EN 374-5:2016
Applications Industrial
AQL 1.5
Color Black
Disposable Single-use
Exterior Finish No Texture
Latex Free Yes
Material Nitrile
Size Small
Thickness (English) 4 mil
Thickness (Metric) 0.10 mm
Model 6112PFS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.