missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Upright Freezers, 50 mm Box Height, with Boxes

ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11254B

Additional Details : Weight : 3.80000kg

 View more versions of this product

Product Code. 12472520

  • £160.00 / Each
Estimated Shipment: 15-07-2024
to see stock.
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Includes standard white cardboard boxes with 9 x 9 cell dividers
  • Holds is expressed by Height x Dept
Specifications

Specifications

White cardboard boxes, 9 x 9 cell dividers
5 x 5
Aluminum
683 x 139 x 276 mm
Seawater resistant
Boxes (25)
Upright Freezers
50 mm (Box)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers