missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ ECO AluRack for Upright Freezers, 100 mm Box Height

ECO AluRack fits into any upright freezer and can be used with all standard cryoboxes.

Brand:  Tenak™ TE11310

Additional Details : Weight : 1.50000kg

 View more versions of this product

Product Code. 12402560

  • £79.15 / Each
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

  • Made of seawater resistant aluminum
  • Fits into any freezer and can be used with all standard cryoboxes
  • Racks can be used with all freezer brands
  • Without white cardboard boxes
  • Holds is expressed by Height x Dept
Specifications

Specifications

Seawater resistant
Boxes (10)
Upright Freezers
100 mm (Box)
2 x 5
Aluminum
683 x 139 x 210 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers