Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 85 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
£54.75 - £72.35
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 85 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12438200
|
Tenak™
TE15210 |
Microtiterplates (3) |
£54.75
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12448200
|
Tenak™
TE15212 |
Microtiterplates (4) |
£57.65
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12458200
|
Tenak™
TE15214 |
Microtiterplates (5) |
£61.60
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12468200
|
Tenak™
TE15216 |
Microtiterplates (6) |
£64.50
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12478200
|
Tenak™
TE15218 |
Microtiterplates (7) |
£67.95
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
12488200
|
Tenak™
TE15220 |
Microtiterplates (8) |
£72.35
Each |
Estimated Shipment: 27-06-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
85 mm |