Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 25 mm Shelf Height
Product Code. 12438180 Shop All Tenak Products
Click to view available options
Holds:
Microtiterplates (11)
Microtiterplates (13)
Microtiterplates (17)
Microtiterplates (19)
Microtiterplates (21)
Microtiterplates (23)
Microtiterplates (27)
Unit Size:
Each
Product Code. 12438180
Brand: Tenak™ TE15146
Only null left
Add to basket
Add to basket
Eco AluRack for chest freezers is designed to hold microtiterplates.
  • For use in chest freezers
  • Racks can either be used as one plate per shelf or the plates can be stacked
  • Constructed of aluminum
Specifications
Material Aluminum
Type Freezer Rack
For Use With (Equipment) Chest Frseezers
Holds Microtiterplates (27)
Dimensions (L x W x H) 135 x 92 x 673 mm
Height (Metric) 25 mm