Learn More
Tenak™ Eco AluRack for Chest Freezer, for Microtiterplates, 16 mm Shelf Height
Eco AluRack for chest freezers is designed to hold microtiterplates.
£78.20 - £136.00
Specifications
Material | Aluminum |
---|---|
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 16 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
12408170
|
Tenak™
TE15108 |
Microtiterplates (29) |
£111.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12418170
|
Tenak™
TE15110 |
Microtiterplates (32) |
£119.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12428170
|
Tenak™
TE15112 |
Microtiterplates (35) |
£128.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12438170
|
Tenak™
TE15114 |
Microtiterplates (39) |
£136.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
12468160
|
Tenak™
TE15100 |
Microtiterplates (16) |
£78.20
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12478160
|
Tenak™
TE15102 |
Microtiterplates (19) |
£86.00
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12488160
|
Tenak™
TE15104 |
Microtiterplates (22) |
£94.30
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
12498160
|
Tenak™
TE15106 |
Microtiterplates (26) |
£103.00
Each |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks can either be used as one plate per shelf or the plates can be stacked
- Constructed of aluminum
Specifications
Aluminum | |
Chest Frseezers | |
16 mm |