missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ ECO 8/ECO 16/ECO 25 Test Tube
Velp Scientifica™ ECO 8/ECO 16/ECO 25 Test Tube constructed of glass with internal diameter of 14 mm and external diameter 16 mm.
Brand: Velp Scientifica™ CM0091680
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Inlcludes 20 glass test tubes
- For use with ECO 8, ECO 16, ECO 25
Specifications
Test Tube | |
Glass | |
14 mm |
ECO 8/ECO 16/ECO25 | |
1 Each | |
16 mm |