Learn More
ECM-1/Secretory Component P85 Antibody (ECM1/792), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47804MFV500
This item is not returnable.
View return policy
Description
ECM-1/Secretory Component P85 Monoclonal antibody specifically detects ECM-1/Secretory Component P85 in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| ECM-1/Secretory Component P85 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| 0.1 mL | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| ECM1/792 | |
| Flow Cytometry | |
| ECM1, extracellular matrix protein 1, Secretory Component Glycoprotein, Secretory component p85 | |
| 1893 | |
| Purified |