missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Eclipse™ Hypodermic Injection Needle with SmartSlip™ Technology
Description
- Unique SmartSlip technology helps to ensure a secure connection to Luer slip syringes
- Bevel oriented to safety cover accommodates low angle injections
- One technology for skin injection and blood collection
- Requires minimal change in injection technique
- The hub incorporates a plastic clip to encourage user to attach needle more securely to Luer slip syringe
- Wide textured finger pad provides safe activation area for thumb or forefinger
- Integral safety cover and extended sidewall to enhance protection
- Needle remains locked inside activated needle cover
- Activates with a touch of a finger – no hard surface required for activation
- Colour coded hub for easy needle size identification
Specifications
Specifications
| Certifications/Compliance | EU directive |
| Color | Pink |
| Needle Gauge | 18 G |
| Diameter (Metric) Outer Needle | 1.2 mm |
| For Use With (Application) | Hypodermic injections |
| Includes | Safety cover |
| Needle Point Style | Bevel |
| For Use With (Equipment) | Luer Slip Syringe |
| Connection Type | Luer |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?