missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 200 μL Siliconized Pipette Tips

Product Code. 17577826
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack, 96
Unit Size:
Pack of 1000
Pack of 960
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format unitSize
17577826 Sterile Rack, 96 Pack of 960
17567826 Non-sterile Rack, 96 Pack of 960
17557826 Non-sterile Bagged Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17577826 Brand Labcon™ Brand No. 10629650069
Only null left
Add to basket
Add to basket

These beveled point, siliconized pipette tips come conveniently packaged in bags or racks.

  • Siliconized for viscous samples
  • Fits most standard 10 μL pipettes
  • Polypropylene USP Class VI Resin
  • Made in USA

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Clear
Rack Color Purple
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 49.7 mm
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Clean Claims ATP, RNase/Dnase, Endotoxin-free
For Use With (Application) Micropipetting
Sterilization Method E-beam
Features Siliconized
Format Rack, 96
Tip Style Rectangle
Quantity 96 Tips/Rack, 10 Racks/Pk., 10 Pk./Cs.
Disposable Single-use
Graduated No
Material Polypropylene Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.