missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 200 μL Graduated Pipette Tips
GREENER_CHOICE

Product Code. 16673952
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Color:
Clear
Natural
Yellow
Format:
Bagged (1000)
Bagged (30,000)
Individually Packaged
Mini Refill
Rack
Rack, 10 x 96
Refill
Stack Rack
Stack Rack Refill
Quantity:
10,000/Cs.
1920/Cs.
2000/Cs.
30,000/Cs.
4800/Cs.
960/Cs.
9600/Cs.
Unit Size:
Pack of 1000
Pack of 1536
Pack of 200
Pack of 30000
Pack of 480
Pack of 960
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Color Format Quantity unitSize
16673952 Non-sterile Clear Refill 9600/Cs. Pack of 960
16683952 Sterile Clear Refill 9600/Cs. Pack of 960
16693952 Non-sterile Clear Bagged (1000) 10,000/Cs. Pack of 1000
16603962 Non-sterile Clear Rack, 10 x 96 9600/Cs. Pack of 960
16613962 Sterile Clear Rack, 10 x 96 9600/Cs. Pack of 960
16671923 Sterile Natural Individually Packaged 2000/Cs. Pack of 200
17577836 Non-sterile Yellow Mini Refill 4800/Cs. Pack of 480
17587836 Non-sterile Clear Stack Rack 1920/Cs. Pack of 960
17597836 Non-sterile Clear Stack Rack Refill 1920/Cs. Pack of 960
17507846 Non-sterile Clear Rack 1920/Cs. Pack of 960
17517846 Sterile Clear Rack 1920/Cs. Pack of 960
17527846 Non-sterile Clear Bagged (30,000) 30,000/Cs. Pack of 30000
17537846 Non-sterile Clear Stack Rack 960/Cs. Pack of 960
17547846 Non-sterile Clear Stack Rack Refill 960/Cs. Pack of 960
17557846 Sterile Clear Stack Rack 960/Cs. Pack of 1536
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Product Code. 16673952 Supplier Labcon™ Supplier No. 10932600009
Only null left
Add to basket
Add to basket

These graduated pipette tips fit most popular 200 μL pipettors.

  • Beveled Point
  • If sterile, Validated Sterilization (ISO 11137) for use in Research and Medical Labs
  • Polypropylene USP Class VI Resin
  • Made in USA

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.95 in.
Length (Metric) 49.7 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase, and Endotoxin-free
For Use With (Application) Liquid Handling
Format Refill
Tip Style Beveled
Packaging 960 Tips/Refill, 10 Refills/Cs.
Quantity 9600/Cs.
ACT Label Yes
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Eclipse
Material Polypropylene Resin
Graduations 10, 20, 50, 100, 200 μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.