missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 200 μL Yellow Pipette Tips

Product Code. 17567796
Change view
Click to view available options
Format:
Bagged
Rack, 192
Rack, 96
Refill
Stack Rack
Sterility:
Non-sterile
Sterile
Unit Size:
Pack of 1000
Pack of 960
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Sterility unitSize
17567796 Stack Rack Sterile Pack of 960
16643892 Bagged Non-sterile Pack of 1000
16653892 Rack, 96 Sterile Pack of 960
16681903 Rack, 96 Non-sterile Pack of 960
17537796 Refill Non-sterile Pack of 960
17547796 Stack Rack Non-sterile Pack of 960
17557796 Rack, 192 Non-sterile Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17567796 Supplier Labcon™ Supplier No. 10327950069
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These pipette tips fit 100 and 200 μL Pipettors.

  • If sterile, Validated Sterilization (ISO 11137) for use in Research and Medical Labs
  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Polypropylene USP Class VI Resin
  • Made in USA

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Sterile
Color Yellow
Autoclavable Autoclavable
Length (English) 1.95 in.
Length (Metric) 49.7 mm
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Clean Claims ATP, RNase/Dnase, Endotoxin-free
For Use With (Application) Liquid Handling
Sterilization Method E-beam
Format Stack Rack
Tip Style Beveled
Quantity 192 Tips/Rack, 5 Racks/Pk., 10 Pk./Cs.
Disposable Single-use
For Use With (Equipment) 100 and 200 μL Pipettors
Graduated No
Product Line Eclipse
Material Polypropylene Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.