missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ Eclipse™ 20 μL Pipette Tips for Rainin™ LTS Pipettors

Product Code. 17527776
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Mini Refill
Rack, 10 x 96
Rack, 192
Refill
Stack Rack
Quantity:
1000 Tips/Bag, 10 Bags/Cs.
1056 Tips/Refill, 10 Refills/Cs.
480 Tips/Mini Refill, 10 Refills/Cs.
92 Tips/Rack, 5 Racks/Pk., 10 Pk./Cs.
960 Tips/Pk., 10 Pk./Cs.
960 Tips/Rack, 10 Racks/Cs.
Unit Size:
Pack of 1000
Pack of 1056
Pack of 480
Pack of 960
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Format Quantity unitSize
17527776 Non-sterile Stack Rack 960 Tips/Rack, 10 Racks/Cs. Pack of 960
16623842 Non-sterile Refill 1056 Tips/Refill, 10 Refills/Cs. Pack of 1056
16633842 Sterile Refill 1056 Tips/Refill, 10 Refills/Cs. Pack of 1056
16643842 Non-sterile Bagged 1000 Tips/Bag, 10 Bags/Cs. Pack of 1000
16653842 Non-sterile Rack, 10 x 96 960 Tips/Pk., 10 Pk./Cs. Pack of 960
16663842 Sterile Rack, 10 x 96 960 Tips/Pk., 10 Pk./Cs. Pack of 960
17517776 Non-sterile Mini Refill 480 Tips/Mini Refill, 10 Refills/Cs. Pack of 480
17537776 Non-sterile Rack, 192 92 Tips/Rack, 5 Racks/Pk., 10 Pk./Cs. Pack of 960
17547776 Sterile Rack, 192 92 Tips/Rack, 5 Racks/Pk., 10 Pk./Cs. Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 17527776 Supplier Labcon™ Supplier No. 10117000069
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These pipette tips have low mounting and ejection forces less than 3 kg.

  • ATP, RNase/DNase and Endotoxin-free (non-pyrogenic)
  • Polypropylene USP Class VI Resin
  • Autoclavable
  • Made in USA

Specifications

Volume (Metric) 20 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.9 in.
Length (Metric) 48.3 mm
Certifications/Compliance ISO 9001:2015, FDA CFR21 GMP, USP Class VI
Clean Claims ATP, RNase/Dnase, Endotoxin-free
For Use With (Application) Liquid Handling
Format Stack Rack
Tip Style UltraFine
Quantity 960 Tips/Rack, 10 Racks/Cs.
ACT Label Yes
Disposable Single-use
For Use With (Equipment) Rainin LTS
Graduated No
Product Line Eclipse
Material Polypropylene Resin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.