missing translation for 'onlineSavingsMsg'
Learn More

Labcon Eclipse™ 10 μL Extra Long Graduated Pipette Tips with UltraFine™ Point
GREENER_CHOICE

Product Code. 16623902
Click to view available options
Sterility:
Non-sterile
Sterile
Format:
Bagged
Rack
Unit Size:
Pack of 1000
Pack of 960

Product Code. 16623902

Brand: Labcon 10378000009

Only null left
Add to basket
Add to basket

Fits Popular 10 μL Pipettors

  • Pipette Tip
  • Eclipse™ Extra Long
  • Graduated at 2 μL and 10 μL
  • With UltraFine™ point
  • Fits Popular 10 μL Pipettors
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Made in USA

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (English) 1.87 in.
Length (Metric) 47.6 mm
Certifications/Compliance ISO 9001:2015; FDA CFR21 GMP
Clean Claims ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
For Use With (Application) Liquid Handing
Format Bagged
Tip Style UltraFine
Packaging 1000 Tips/Bag, 10 Bags/Cs.
Quantity 1000 tips/bag
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line Eclipse
Material Polypropylene Resin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.