missing translation for 'onlineSavingsMsg'
Learn More

ECHS1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18409739 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409739 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409739 Supplier Novus Biologicals Supplier No. NBP296977JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ECHS1 Polyclonal antibody specifically detects ECHS1 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECHS1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias EC 4.2.1, enoyl CoA hydratase, short chain, 1, mitochondrial, enoyl Coenzyme A hydratase, short chain, 1, mitochondrial, Enoyl-CoA hydratase 1, enoyl-CoA hydratase, mitochondrial, SCEHEC 4.2.1.17, Short-chain enoyl-CoA hydratase
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ECHS1 (rh ECHS1; AAH08906.1; 28A-290Q)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1892
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.