missing translation for 'onlineSavingsMsg'
Learn More

Echothiophate Iodide, TRC

Product Code. 30385630
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385630 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385630 Supplier Toronto Research Chemicals Supplier No. TRCE3258001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Echothiophate iodide
CAS 513-10-0
Molecular Formula C9 H23 N O3 P S . I
Quantity 1 g
Synonym Ethanaminium, 2-[(diethoxyphosphinyl)thio]-N,N,N-trimethyl-, iodide (1:1), (2-Mercaptoethyl)trimethylammonium iodide, O,O-diethyl phosphorothioate (6CI), (2-Mercaptoethyl)trimethylammonium iodide, S-ester with O,O-diethyl phosphorothioate (7CI), Ammonium, (2-mercaptoethyl)trimethyl-, iodide, S-ester with O,O-diethyl phosphorothioate (8CI), Ethanaminium, 2-[(diethoxyphosphinyl)thio]-N,N,N-trimethyl-, iodide (9CI), 217MI, Diethoxyphosphinylthiocholine iodide, Diethylphosphorylthiocholine iodide, Echothiopate iodide, Echothiophate iodide, Ecostigmine iodide, Ecostigmini jodidum, Ecothiopate iodide, N-[2-(Diethoxyphosphinylthio)ethyl]trimethylammonium iodide, Phospholine iodide, S-(2-Dimethylaminoethyl) O,O-diethylphosphorothioate methiodide, S-(Diethyl phosphono)thiocholine iodide
InChI Formula InChI=1S/C9H23NO3PS.HI/c1-6-12-14(11,13-7-2)15-9-8-10(3,4)5;/h6-9H2,1-5H3;1H/q+1;/p-1
SMILES [I-].CCOP(=O)(OCC)SCC[N+](C)(C)C
IUPAC Name 2-diethoxyphosphorylsulfanylethyl(trimethyl)azanium;iodide
Molecular Weight (g/mol) 383.23
Formula Weight 383.0181
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.