missing translation for 'onlineSavingsMsg'
Learn More

Echinacoside, MedChemExpress

Product Code. 16491268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491268 5 mg 5mg
16484727 50 mg 50mg
16448828 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491268 Supplier Medchem Express Supplier No. HYN0020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Echinacoside, one of the phenylethanoids isolated from the stems of Cistanche salsa, effectively inhibits Wnt/β-catenin signaling. Echinacoside elicits neuroprotection by activating Trk receptors and their downstream signal pathways. Antiosteoporotic activity.

Echinacoside, one of the phenylethanoids isolated from the stems of Cistanche salsa, effectively inhibits Wnt/β-catenin signaling. Echinacoside elicits neuroprotection by activating Trk receptors and their downstream signal pathways. Antiosteoporotic activity.

Specifications

Chemical Name or Material Echinacoside
CAS 82854-37-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C35H46O20
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (63.55 mM; Need ultrasonic) ∣H2O : 35.71 mg/mL (45.39 mM; Need ultrasonic)
SMILES OC1=CC(/C=C/C(O[C@H]2[C@H](O[C@H]3[C@H](O)[C@H](O)[C@@H](O)[C@H](C)O3)[C@@H](O)[C@H](OCCC4=CC(O)=C(O)C=C4)O[C@@H]2CO[C@@H]5O[C@H](CO)[C@@H](O)[C@H](O)[C@H]5O)=O)=CC=C1O
Molecular Weight (g/mol) 786.73
Formula Weight 786.73
Percent Purity 98.18%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.