missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECHDC2 Polyclonal Antibody

ECHDC2 Polyclonal Antibody
Brand: Thermo Scientific PA555413
This item is not returnable.
View return policy
Description
ECHDC2 Polyclonal Antibody for IHC
Specifications
| ECHDC2 | |
| Polyclonal | |
| Unconjugated | |
| Echdc2 | |
| 1300017C12Rik; 2610009M20Rik; D4Ertd765e; ECHDC2; enoyl CoA hydratase domain containing 2; enoyl Coenzyme A hydratase domain containing 2; enoyl-CoA hydratase domain containing 2; enoyl-CoA hydratase domain-containing protein 2, mitochondrial; hypothetical protein LOC513795; id:ibd1032; RGD1308525; Unknown (protein for MGC:85763); wu:fc83b02; zgc:56321; zgc:85763 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 55268 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86YB7 | |
| Echdc2 | |
| Recombinant protein corresponding to Human ECHDC2. Recombinant protein control fragment (Product #RP-94152). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction