missing translation for 'onlineSavingsMsg'
Learn More

ECH1 Antibody, R&D Systems™

Product Code. 18604866 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18604866 50 μg 50µL
18659517 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18604866 Supplier Novus Biologicals Supplier No. NBP30032450ul
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

ECH1 Polyclonal specifically detects ECH1 in Human samples. It is validated for ELISA/Immunohistochemistry/Immunohistochemistry (Paraffin)/Immunoprecipitation/Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ECH1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 0.5-2 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2500-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias delta(3,5)-Delta(2,4)-dienoyl-CoA isomerase, mitochondrial, delta3,5-delta2,4-dienoyl-CoA isomerase, dienoyl-CoA isomerase, EC 5.3.3.-, enoyl CoA hydratase 1, peroxisomal, enoyl Coenzyme A hydratase 1, peroxisomal, HPXEL, peroxisomal enoyl-CoA hydratase 1
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human ECH1 (Uniprot#: Q13011; Thr34-Leu328)
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 1891
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.