Learn More
ECE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15418-1-AP-150UL
 View more versions of this product
Product Code. 16804584
  • £349.00 / 150µL
Description
Converts big endothelin-1 to endothelin-1. Also involved in the processing of various neuroendocrine peptides, including neurotensin, angiotensin I, substance P, proenkephalin-derived peptides, and prodynorphin-derived peptides. May limit beta-amyloid peptide accumulation in brain. May also have methyltransferase activity.
Specifications
Specifications
ECE2
Polyclonal
Unconjugated
ECE2
ECE2, endothelin converting enzyme 2, KIAA0604
Rabbit
Antigen Affinity Chromatography
RUO
107522, 408243, 9718
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O60344, Q80Z60
Ece2
ECE2 Fusion Protein Ag7706
150 μL
Primary
Human, Mouse, Rat