missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ECE-2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05677-20ul
This item is not returnable.
View return policy
Description
ECE-2 Polyclonal antibody specifically detects ECE-2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| ECE-2 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000 | |
| EC 3.4.24, EC 3.4.24.71, ECE-2, endothelin converting enzyme 2, endothelin-converting enzyme 2, KIAA0604MGC2408, MGC17664, MGC78487 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human ECE-2 (NP_115707.2). MASPGAGRAPPELPERNCGYREVEYWDQRYQGAADSAPYDWFGDFSSFRALLEPELRPEDRILVLGCGNSALSYELFLGGFPNVTSVDYSSVVVAAMQARHAHVPQLRWETMDVRKLDFPSASFDVVLEKGTLDALLAGERDPWTVSSEGVHTVDQVLSE | |
| 20 μg | |
| Signal Transduction | |
| 9718 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction