missing translation for 'onlineSavingsMsg'
Learn More

Ecdysterone

Product Code. 15465359
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465359 50 mg 50mg
15455359 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15465359 Supplier Thermo Scientific Alfa Aesar Supplier No. J63091.MF
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

An insect steroid hormone that induces apoptosis | CAS: 5289-74-7 | C27H44O7 | 480.64 g/mol

Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ecdysterone is an insect steroid hormone that induces apoptosis, cell proliferation, growth by controlling gene expression involved in molting and metamorphosis. And it is also used as a ecdysteroid in both plant and animal species that controls cell death.

Solubility
Soluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5289-74-7
Molecular Formula C27H44O7
Molecular Weight (g/mol) 480.64
MDL Number MFCD00036740
InChI Key NKDFYOWSKOHCCO-YPVLXUMRSA-N
Synonym 20-hydroxyecdysone, ecdysterone, beta-ecdysone, crustecdysone, 20-oh ecdysone, the-7, polypodine a, insect moulting hormone, commisterone, crustecdyson
PubChem CID 5459840
ChEBI CHEBI:16587
IUPAC Name (1S,3aS,5aR,7R,8S,9aR,9bR,11aR)-3a,7,8-trihydroxy-9a,11a-dimethyl-1-[(2R,3R)-2,3,6-trihydroxy-6-methylheptan-2-yl]-1H,2H,3H,3aH,5H,5aH,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-5-one
SMILES CC(C)(O)CC[C@@H](O)[C@](C)(O)[C@H]1CC[C@@]2(O)C3=CC(=O)[C@@H]4C[C@@H](O)[C@@H](O)C[C@]4(C)[C@H]3CC[C@]12C

Specifications

Melting Point 242°C to 244°C
Color White
Quantity 50 mg
Beilstein 1917578
Merck Index 14,3491
Solubility Information Soluble in alcohol
Formula Weight 480.64
Physical Form Powder
Chemical Name or Material Ecdysterone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.