missing translation for 'onlineSavingsMsg'
Learn More

Ecamsule, TRC

Product Code. 30385920
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385920 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385920 Supplier Toronto Research Chemicals Supplier No. TRCE320700250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ecamsule Solution
CAS 92761-26-7
Purity Grade Notes HPLC
Molecular Formula C28 H34 O8 S2
Quantity 250 mg
Synonym 3,3'-(1,4-Phenylenedimethylidyne)bis[7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonic acid], 1,4-Di(2-oxo-10-Sulfo-3-bornylidenemethyl)Benzene, 1,4-Di(2-oxo-10-sulpho-3-bornylidenemethyl)benzene, 3,3'-(1,4-Phenylenedimethine)bis(7,7-dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic acid, Ecamsule, Mexoryl SX, Terephthalylidene-3,3'-dicamphor-10,10'-disulfonic acid
InChI Formula InChI=1S/C28H34O8S2/c1-25(2)21-9-11-27(25,15-37(31,32)33)23(29)19(21)13-17-5-7-18(8-6-17)14-20-22-10-12-28(24(20)30,26(22,3)4)16-38(34,35)36/h5-8,13-14,21-22H,9-12,15-16H2,1-4H3,(H,31,32,33)(H,34,35,36)/b19-13-,20-14-
SMILES CC1(C)C2CCC1(CS(=O)(=O)O)C(=O)/C/2=C\c3ccc(\C=C/4\C5CCC(CS(=O)(=O)O)(C4=O)C5(C)C)cc3
IUPAC Name [(2Z)-2-[[4-[(Z)-[7,7-dimethyl-3-oxo-4-(sulfomethyl)-2-bicyclo[2.2.1]heptanylidene]methyl]phenyl]methylidene]-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid
Molecular Weight (g/mol) 562.6948
Formula Weight 562.1695
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.