missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ecamsule, TRC
Specifications
Specifications
| Chemical Name or Material | Ecamsule Solution |
| CAS | 92761-26-7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28 H34 O8 S2 |
| Quantity | 10 mg |
| Synonym | 3,3'-(1,4-Phenylenedimethylidyne)bis[7,7-dimethyl-2-oxobicyclo[2.2.1]heptane-1-methanesulfonic acid], 1,4-Di(2-oxo-10-Sulfo-3-bornylidenemethyl)Benzene, 1,4-Di(2-oxo-10-sulpho-3-bornylidenemethyl)benzene, 3,3'-(1,4-Phenylenedimethine)bis(7,7-dimethyl-2-oxo-bicyclo[2.2.1]heptane-1-methanesulfonic acid, Ecamsule, Mexoryl SX, Terephthalylidene-3,3'-dicamphor-10,10'-disulfonic acid |
| InChI Formula | InChI=1S/C28H34O8S2/c1-25(2)21-9-11-27(25,15-37(31,32)33)23(29)19(21)13-17-5-7-18(8-6-17)14-20-22-10-12-28(24(20)30,26(22,3)4)16-38(34,35)36/h5-8,13-14,21-22H,9-12,15-16H2,1-4H3,(H,31,32,33)(H,34,35,36)/b19-13-,20-14- |
| SMILES | CC1(C)C2CCC1(CS(=O)(=O)O)C(=O)/C/2=C\c3ccc(\C=C/4\C5CCC(CS(=O)(=O)O)(C4=O)C5(C)C)cc3 |
| IUPAC Name | [(2Z)-2-[[4-[(Z)-[7,7-dimethyl-3-oxo-4-(sulfomethyl)-2-bicyclo[2.2.1]heptanylidene]methyl]phenyl]methylidene]-7,7-dimethyl-3-oxo-4-bicyclo[2.2.1]heptanyl]methanesulfonic acid |
| Molecular Weight (g/mol) | 562.6948 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?