missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies EC23

Product Code. 17140172
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17140172 1 mg 1mg
17150172 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17140172 Supplier STEMCELL Technologies Supplier No. 73102

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Retinoid pathway activator; Activates retinoic acid receptor (RAR)

  • EC23 is a retinoic acid receptor (RAR) agonist with pan-RAR activity (EC50: RARα 41 nM, RARβ 0.5 nM, RARγ 0.4 nM), while having no appreciable activity for retinoid X receptors (RXR; EC50> 10μM for all; Gambone et al.) It is a photostable synthetic analog of all-trans retinoic acid (ATRA; Christie et al. 2008)
  • EC23 also weakly activates aryl hydrocarbon receptor (Gambone et al.)
  • Induces neural differentiation of human pluripotent stem cells, similarly to ATRA (Christie et al. 2010; Clemens et al.)
  • Induces neuronal differentiation of human fetal neural progenitor cell line ReNcell 197VM (Christie et al. 2010)

Specifications

CAS 104561-41-3
Synonym AGN 190205; BASF 46928
Quantity 1 mg
Molecular Formula C23H24O2
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.