Learn More
Macherey Nagel EC HPLC column EC 150/4NUCLEODUR HILIC, 3 µm length: 150 mm,
Brand:  Macherey Nagel 760533.40
EC HPLC column EC 150/4NUCLEODUR HILIC, 3 µm length: 150 mm,
Product Code. 12788862
  • £718.00 / Pack of 1