Learn More
Macherey Nagel EC HPLC column EC 150/2NUCLEOSHELL PFP, 2.7 µm length: 150 mm,
Brand:  Macherey Nagel 763536.20
EC HPLC column EC 150/2NUCLEOSHELL PFP, 2.7 µm length: 150 mm,
Product Code. 12793551
  • £733.00 / Pack of 1