Learn More
Macherey Nagel EC HPLC column EC 100/4NUCLEODUR C18 HTec, 5 µm length: 100 mm,
Brand:  Macherey Nagel 760313.40
EC HPLC column EC 100/4NUCLEODUR C18 HTec, 5 µm length: 100 mm,
Product Code. 12793431
  • £576.00 / Pack of 1
Only null left
Estimated Shipment: 11-06-2026