missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ EC 23

Product Code. 18792644
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18792644 50 mg 50mg
18752694 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18792644 Supplier Tocris Bioscience™ Supplier No. 4011/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Synthetic retinoid; induces differentiation of stem cells

Photostable synthetic retinoid; analog of ATRA (Cat. No. 0695). Displays similar activity to ATRA when tested on the embryonal carcinoma stem cell model TERA2.cl.SP12. Potently induces neural differentiation in human pluripotent embryonic stem cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104561-41-3
Molecular Formula C23H24O2

Specifications

Quantity 50 mg
Formula Weight 332.44
Purity >99%
Molecular Weight (g/mol) 332.443
Chemical Name or Material EC 23
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.