missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBA Lifesciences™ EBV LMP2 Peptide CLGGLLTMV (HLA-A*0201)
Shop All IBA Lifesciences Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
EBV LMP2 peptide CLGGLLTMV (HLA-A*0201) is a single peptide for stimulation of T cells. The peptide from human Epstein-Barr virus (EBV) latent membrane protein 2 (LMP2) is synthesized as it is presented by the MHC class I HLA-A*0201 allele and can be used for targeted in vitro generation and expansion of antigen-specific CD8+ T cells.
- Peptides have been purified by HPLC using a water and acetonitrile gradient
- Endotoxin-free
Specifications
Specifications
| Purity | >85% |
| Endotoxin Level | Endotoxin-free |
| For Use With (Application) | T Cell Stimulation |
| Lyophilized | Yes |
| Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction