missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EBV Early Antigens Monoclonal antibody specifically detects EBV Early Antigens in Virus samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | EBV Early Antigens |
| Applications | Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1108-1 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/ Immunofluorescence 1-2 ug/mL |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | Chronic ulcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?