missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ebselen

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Ebselen is a glutathione peroxidase mimic; peroxynitrite scavenger. Inhibits ferroptosis. Also inhibits lipoxygenase, cyclooxygenase, nitric oxide synthase, protein kinase C and H+/K+-ATPase activity. Inhibits the hepatic carcinogenic effects of aflatoxin B1. Antioxidant and anti-inflammatory. Selectively inhibits Gq protein signaling and enhances differentiation of brown adipocytes. Inhibits SARS-CoV-2 Mpro in vitro (IC50 = 0.67 μM in FRET assay)
Specifications
Specifications
| CAS | 60940-34-3 |
| Quantity | 10 mg |
| Molecular Formula | 2-Phenyl-1,2-benzisoselenazol-3(2 H)-one |
| Product Type | Ebselen |
| Purity | 98% |
| Molecular Weight (g/mol) | M.W. (Observed): 274.18 |
| Inhibitors | Ebselen |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction