missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ebselen
GREENER_CHOICE

Product Code. 18767426
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18767426

Brand: Tocris Bioscience™ 5245/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glutathione peroxidase mimic; peroxynitrite scavenger

Ebselen is a glutathione peroxidase mimic; peroxynitrite scavenger. Inhibits ferroptosis. Also inhibits lipoxygenase, cyclooxygenase, nitric oxide synthase, protein kinase C and H+/K+-ATPase activity. Inhibits the hepatic carcinogenic effects of aflatoxin B1. Antioxidant and anti-inflammatory. Selectively inhibits Gq protein signaling and enhances differentiation of brown adipocytes. Inhibits SARS-CoV-2 Mpro in vitro (IC50 = 0.67 μM in FRET assay)
TRUSTED_SUSTAINABILITY

Specifications

CAS 60940-34-3
Quantity 50 mg
Molecular Formula 2-Phenyl-1,2-benzisoselenazol-3(2 H)-one
Product Type Ebselen
Purity 98%
Molecular Weight (g/mol) M.W. (Observed): 274.18
Inhibitors Ebselen

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.