missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBPL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA562080
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: WQSWLELKKM HQKETSSVKK FQ Highest antigen sequence identity to the following orthologs: Mouse - 50%, Rat - 50%.
EBPL does not possess sterol isomerase activity and does not bind sigma ligands.Specifications
EBPL | |
Polyclonal | |
Unconjugated | |
EBPL | |
EBPL; EBRP; emopamil binding protein like; emopamil binding protein-like; emopamil binding related protein, delta8-delta7 sterol isomerase related protein; emopamil-binding protein-like; Emopamil-binding-related protein; ERP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84650 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BY08 | |
EBPL | |
Recombinant protein corresponding to Human EBPL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only