missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBP50 Recombinant Rabbit Monoclonal Antibody (24GB1270)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556946
This item is not returnable.
View return policy
Description
EBP50 Recombinant Monoclonal Antibody for Western Blot, Flow
NHERF1 (Na+/H+ exchanger regulatory factor 1), also known as EBP50 (ezrin, radixin, moesin-binding phosphoprotein 50) is an adaptor protein, which associates with beta-catenin and is required for its localization at the cell-cell junctions, interacts with various G protein-coupled receptors and regulates their traffic, as well as sodium-hydrogen exchange and sodium-dependent phosphate transport. NHERF1/EBP50 inhibits cell motility and is required to suppress anchorage-independent growth. It contains C-terminal ERM (ezrin, radixin, moesin)-binding region and two N-terminal PDZ (postsynaptic-density-95/disc-large/ZO1 homology) domains and is able to form head-to-tail intramolecular conformation to regulate its interactions.
Specifications
| EBP50 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SLC9A3R1 | |
| EBP50; EBP-50; ERM-binding phosphoprotein; ezrin-radixin-moesin binding phosphoprotein-50; ezrin-radixin-moesin-binding phosphoprotein 50; Na(+)/H(+) exchange regulatory cofactor NHE-RF1; Na+/H+ exchange regulatory co-factor; Nherf; NHE-RF; Nherf1; NHERF-1; NPHLOP2; Regulatory cofactor of Na(+)/H(+) exchanger; SLC9A3 regulator 1; Slc9a3r1; So; Sodium-hydrogen exchanger regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulator 1; solute carrier family 9 (sodium/hydrogen exchanger), isoform 3 regulatory factor 1; solute carrier family 9 (sodium/hydrogen exchanger), member 3 regulator 1; solute carrier family 9 isoform A3 regulatory factor 1; solute carrier family 9, subfamily A (NHE3, cation proton antiporter 3), member 3 regulator 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 59114, 9368 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB1270 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O14745, Q9JJ19 | |
| SLC9A3R1 | |
| A synthesized peptide derived from human EBP50 (aa 51-120). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction