Learn More
Invitrogen™ EBP1 Recombinant Rabbit Monoclonal Antibody (24GB580)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557634
 View more versions of this product
Product Code. 30276536
  • £488.00 / 100µL
Description
EBP1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EBP1
Recombinant Monoclonal
Unconjugated
Pa2g4
38kDa; AA672939; Cell cycle protein p38-2G4 homolog; Ebp1; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4 1; hG4-1; IRES-specific cellular trans-acting factor 45 kDa; ITAF45; MGC81621; MGC94070; Mpp1; p38 2G4; p38-2G4; Pa2g4; Plfap; proliferation-associated 2G4; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; proliferation-associated protein 1; Proliferation-associated protein 2G4; protein p38-2G4; si:dz150i12.2; wu:fb19b11; wu:ft56d05; zgc:86732
Rabbit
Affinity chromatography
RUO
18813, 288778, 5036
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB580
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P50580, Q6AYD3, Q9UQ80
Pa2g4
A synthesized peptide derived from human EBP1 / PA2G4 (aa 300-394).
100 μL
Primary
Human, Mouse, Rat