missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBP1 Polyclonal Antibody

Description
EBP1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
This gene encodes an RNA-binding protein that is involved in growth regulation. This protein is present in pre-ribosomal ribonucleoprotein complexes and may be involved in ribosome assembly and the regulation of intermediate and late steps of rRNA processing. This protein can interact with the cytoplasmic domain of the ErbB3 receptor and may contribute to transducing growth regulatory signals. This protein is also a transcriptional co-repressor of androgen receptor-regulated genes and other cell cycle regulatory genes through its interactions with histone deacetylases. This protein has been implicated in growth inhibition and the induction of differentiation of human cancer cells. Six pseudogenes, located on chromosomes 3, 6, 9, 18, 20 and X, have been identified.
Specifications
Specifications
| Antigen | EBP1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 |
| Gene | Pa2g4 |
| Gene Accession No. | Q9UQ80 |
| Gene Alias | 38kDa; AA672939; Cell cycle protein p38-2G4 homolog; Ebp1; ErbB-3 binding protein 1; ErbB3-binding protein 1; ErbB3-binding protein Ebp1; hG4 1; hG4-1; IRES-specific cellular trans-acting factor 45 kDa; ITAF45; MGC81621; MGC94070; Mpp1; p38 2G4; p38-2G4; Pa2g4; Plfap; proliferation-associated 2G4; proliferation-associated 2G4, 38kD; proliferation-associated 2G4, 38kDa; proliferation-associated protein 1; Proliferation-associated protein 2G4; protein p38-2G4; si:dz150i12.2; wu:fb19b11; wu:ft56d05; zgc:86732 |
| Gene Symbols | Pa2g4 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?