missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5107117
This item is not returnable.
View return policy
Description
Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 85%; Pig: 93%; Rabbit: 93%; Rat: 85%.
EBP is an integral membrane protein of the endoplasmic reticulum. It is a high affinity binding protein for the antiischemic phenylalkylamine Ca2+ antagonist [3H]emopamil and the photoaffinity label [3H]azidopamil. It is similar to sigma receptors and may be a member of a superfamily of high affinity drug-binding proteins in the endoplasmic reticulum of different tissues. These aromatic amino acid residues have been suggested to be involved in the drug transport by the P-glycoprotein. Mutations in EBP gene cause Chondrodysplasia punctata 2 (CDPX2; also known as Conradi-Hunermann syndrome).Emopamil-binding protein (EBP) is an integral membrane protein of the endoplasmic reticulum. It is a high affinity binding protein for the antiischemic phenylalkylamine Ca2+ antagonist [3H]emopamil and the photoaffinity label [3H]azidopamil. It is similar to sigma receptors and may be a member of a superfamily of high affinity drug-binding proteins in the endoplasmic reticulum of different tissues. EBP shares structural features with bacterial and eukaryontic drug transporting proteins. It has four putative transmembrane segments and contains two conserved glutamate residues which may be involved in the transport of cationic amphiphilics. Another prominent feature of EBP is its high content of aromatic amino acid residues (>23%) in its transmembrane segments. These aromatic amino acid residues have been suggested to be involved in the drug transport by the P-glycoprotein. Mutations in this gene cause Chondrodysplasia punctata 2 (CDPX2; also known as Conradi-Hunermann syndrome).
Specifications
| EBP | |
| Polyclonal | |
| Unconjugated | |
| EBP | |
| 3-beta-hydroxysteroid-Delta(8),Delta(7)-isomerase; 3-beta-hydroxysteroid-delta-8,delta-7-isomerase; AI255399; CDPX2; CHO2; cholestenol Delta-isomerase; Chondrodysplasia punctata-2, X-linked dominant (Happle syndrome); CPX; CPXD; D8-D7 sterol isomerase; delta(8)-Delta(7) sterol isomerase; Ebp; emopamil binding protein (sterol isomerase); emopamil-binding protein; MEND; Msi; Pabp; phenylalkylamine Ca2+ antagonist (emopamil) binding protein; SI; sterol 8-isomerase; Td | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10682 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q15125 | |
| EBP | |
| Synthetic peptide directed towards the N terminal region of human EBP. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction