missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06227CL1
This item is not returnable.
View return policy
Description
EBP Polyclonal antibody specifically detects EBP in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| EBP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CoraFluor 1 | |
| 3-beta-hydroxysteroid-Delta(8), CDPX2, Cholestenol Delta-isomerase, CPX3-beta-hydroxysteroid-delta-8, CPXDX-linked dominant (Happle syndrome), D8-D7 sterol isomerase, Delta(7)-isomerase, Delta(8)-Delta(7) sterol isomerase, delta-7-isomerase, EC 5.3.3.5, emopamil binding protein (sterol isomerase), Emopamil-binding protein, sterol 8-isomerase | |
| Produced in rabbits immunized with E. coli-derived Human EBP fragment. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 10682 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction