missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EBOV VP35 protein Polyclonal antibody specifically detects EBOV VP35 protein in Virus samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | EBOV VP35 protein |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 1% BSA, 20% Glycerol (pH7) |
| Gene Alias | Capsid protein VP35, Ebola VP35, eVP35, Polymerase cofactor VP35, Viral protein 35 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of EBOV VP35 protein protein (Zaire ebolavirus (strain Mayinga-76)). The exact sequence is proprietary. |
| Purification Method | Antigen Affinity-purified |
| Quantity | 100 μg |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?