missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
EBOV GP2 Monoclonal antibody specifically detects EBOV GP2 in Virus samples. It is validated for ELISA
Specifications
Specifications
| Classification | Monoclonal |
| Host Species | Mouse |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 3160774 |
| Content And Storage | Store at 4C in the dark. |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?