missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBOV GP1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05820-100ul
This item is not returnable.
View return policy
Description
EBOV GP1 Polyclonal antibody specifically detects EBOV GP1 in Ebola virus samples. It is validated for Western Blot
Specifications
| EBOV GP1 | |
| Polyclonal | |
| Western Blot 1:1000-1:5000 | |
| second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein | |
| Produced in rabbits immunized with purified, recombinant EBOV GP1 (Accession#: ACI28624.1; Met1-Arg501). | |
| 100 μg | |
| Primary | |
| Ebola virus | |
| Purified |
| Western Blot | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 3160774 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction