missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EBNA1BP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA545616
This item is not returnable.
View return policy
Description
Peptide sequence: VTLGPVPEIG GSEAPAPQNK DQKAVDPEDD FQREMSFYRQ AQAAVLAVLP Sequence homology: Human: 100%
EBNA1BP2 Polyclonal antibody specifically detects EBNA1BP2 in Human samples. It is validated for Western Blot.
Specifications
| EBNA1BP2 | |
| Polyclonal | |
| Unconjugated | |
| EBNA1BP2 | |
| 1810014B19Rik; AA537058; B830003A16Rik; C81321; EBNA1 binding protein 2; EBNA1-binding protein 2; Ebna1bp2; EBP2; NOBP; nuclear FGF3 binding protein; nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; p40; Probable rRNA-processing protein EBP2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10969 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q99848 | |
| EBNA1BP2 | |
| Synthetic peptide directed towards the middle region of human EBNA1BP2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction