missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EBNA1BP2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555326
This item is not returnable.
View return policy
Description
Immunogen sequence: ESDESLVTDR ELQDAFSRGL LKPGLNVVLE GPKKAVNDVN GLKQCLAEFK RDLEWVERLD VTLGPVPEIG GSEAPAPQNK DQKAVDPEDD Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
EBNA1BP2 is required for the processing of the 27S pre-rRNA.
Specifications
| EBNA1BP2 | |
| Polyclonal | |
| Unconjugated | |
| EBNA1BP2 | |
| 1810014B19Rik; AA537058; B830003A16Rik; C81321; EBNA1 binding protein 2; EBNA1-binding protein 2; Ebna1bp2; EBP2; NOBP; nuclear FGF3 binding protein; nucleolar protein p40; nucleolar protein p40; homolog of yeast EBNA1-binding protein; p40; Probable rRNA-processing protein EBP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10969 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99848 | |
| EBNA1BP2 | |
| Recombinant protein corresponding to Human EBNA1BP2. Recombinant protein control fragment (Product #RP-94115). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido