missing translation for 'onlineSavingsMsg'
Learn More

EBNA1BP2 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £359.00

Specifications

Antigen EBNA1BP2
Dilution Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18653422
View Documents
Novus Biologicals
NBP2-92575-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18670412
View Documents
Novus Biologicals
NBP2-92575-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EBNA1BP2 Polyclonal antibody specifically detects EBNA1BP2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

EBNA1BP2
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Core ESC Like Genes, Stem Cell Markers
PBS with 50% glycerol, pH7.3.
10969
IgG
Affinity purified
Western Blot 1:1000 - 1:2000, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200
Polyclonal
Purified
RUO
Human, Mouse, Rat
EBNA1 binding protein 2, EBNA1-binding protein 2nucleolar protein p40; homolog of yeast EBNA1-binding protein, EBP2nuclear FGF3 binding protein, NOBP, Nucleolar protein p40, P40, probable rRNA-processing protein EBP2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-306 of human EBNA1BP2 (NP_006815.2). MDTPPLSDSESESDESLVTDRELQDAFSRGLLKPGLNVVLEGPKKAVNDVNGLKQCLAEFKRDLEWVERLDVTLGPVPEIGGSEAPAPQNKDQKAVDPEDDFQREMSFYRQAQAAVLAVLPRLHQLKVPTKRPTDYFAEMAKSDLQMQKIRQKLQTKQAAMERSEKAKQLRALRKYGKKVQTEVLQKRQQEKAHMMNAIKKYQKGFSDKLDFLEGDQKPLAQRKKAGAKGQQMRKGPSAKRRYKNQKFGFGGKKKGSKWNTRESYDDVSSFRAKTAHGRGLKRPGKKGSNKRPGKRTREKMKNRTH
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.